Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Polyethyleneimine, branched, M.W. 600, 99%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Solubility Information | Soluble in toluene,methanol,ethanol,chloroform and xylene. Insoluble in water,alcohol,benzene,ethyl ether and acetone. |
Physical Form | Liquid |
Percent Purity | 99% |
Odor | Amine-like |
Color | Yellow |
Quantity | 500g |
Chemical Name or Material | Polyethyleneimine, branched, M.W. 600 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Hazard Class: 9; Packaging Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures