Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, 20-40 mesh

For the separation of nucleotide mixtures

Manufacturer: Alfa Aesar™ 43302AD

Quantity 50mL

 View more versions of this product

Catalog No. AA43302AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9002-98-6
    MDL Number MFCD00084427
    Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin

    Specifications

    TSCA TSCA
    Physical Form Beads
    Mesh Size 20 to 40 Mesh
    MDL Number MFCD00084427
    Sensitivity Hygroscopic
    CAS 9002-98-6
    Quantity 50mL
    Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit