Learn More
Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, 40-200 mesh
For the separation of nucleotide mixtures
Manufacturer:  Alfa Aesar™ 43303AD
Quantity 50mL
 View more versions of this product
Catalog No. AA43303AD
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
9002-98-6
MFCD00084427
Polyethyleneimine on silica beads anion exchange resin
Specifications
9002-98-6
MFCD00084427
Hygroscopic
Polyethyleneimine on silica beads anion exchange resin
Powder
40 to 200 Mesh
50mL
Safety and Handling
SDS & Safety Documents
TSCA : TSCA