Learn More
Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, benzylated, 40-200 mesh
Solid phase organic synthesis, affinity chromatography, catalyst support
Manufacturer:  Alfa Aesar™ 43305AK
Quantity 250mL
 View more versions of this product
Catalog No. AA43305AK
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
9002-98-6
MFCD00084427
Polyethyleneimine on silica beads anion exchange resin, Benzylated
Specifications
9002-98-6
MFCD00084427
Hygroscopic
Polyethyleneimine on silica beads anion exchange resin, Benzylated
Powder
40 to 200 Mesh
250mL