Alfa Aesar™ Polyimide film, 0.025mm thick x 20mm width 33m

Manufacturer: Alfa Aesar™ 41898UL

Quantity 33m

 View more versions of this product

Catalog No. AA41898UL

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62929-02-6
    MDL Number MFCD00677744
    Chemical Name or Material Polyimide, 0.025mm thick x 20mm width

    Specifications

    Physical Form Film
    MDL Number MFCD00677744
    CAS 62929-02-6
    Quantity 33m
    Chemical Name or Material Polyimide, 0.025mm thick x 20mm width

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit