Learn More
Polymedco™ OC-Light Complete Manual iFOBT Kit
A complete, quick and easy screening method for detection of fecal occult blood
Manufacturer:  POLYMEDCO CANCER DIAGNOSTIC PRODUCTS LLC FOB50
 View more versions of this product
Catalog No. 21-000-100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Waived
Sampling bottles, collection Papers and test strips
5 min.
Detection of Human hemoglobin in feces
50 Pack
Complete manual iFOBT Kit