Learn More
Polymedco™ Sediplast™ Autozero ESR System Available on GSA/VA Contract for Federal Government customers only.
Closed system with unique vented cap: precisely calibrated hole stops the column of blood at true “zero”
Manufacturer:  Polymedco™ S1001
 View more versions of this product
Catalog No. 02-681-7
  Check Availability
Add to cart
Description
Description
  • Pierceable stopper manufactured from a special formulation of linear low-density polyethylene to ensure a leakproof seal during the procedure
  • When the pipet is inserted into the vial, the diaphragm is designed to break away and the unit becomes a completely sealed system
  • Eliminates all manipulation “techniques” in setting sample at the zero mark
  • All excess blood flows into a closed, separate reservoir compartment automatically
  • Includes a stoppered, pre-filled vial containing 0.2mL of 3.8% sodium citrate diluent
  • Simply add 0.8mL of blood and mix
  • Eliminates separate diluent preparation, dilution, and handling steps required in other systems
  • Also available without diluent
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
SEDIPLAST Autozero ESR System
Whole EDTA Blood
Used for hematology applications
Westergren tubes without diluent