Polymedco™ Sediplast™ Autozero ESR System
Closed system with unique vented cap: precisely calibrated hole stops the column of blood at true “zero”
Manufacturer: PolymedcoSupplier Diversity Partner S1080
 View more versions of this product
Catalog No. 23-307374
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With (Equipment) SEDIPLAST Autozero ESR System
    For Use With (Application) Used for hematology applications
    Product Type SediPlast/Wintrobe tube
  • Description & Specifications
Related Products
Product Content Feedback