Learn More
Polymedco™ Venoplast™ Vacuum and Nonvacuum ESR Tubes
SDP
For use with Polymedco Sediten 10-Minute ESR Reader
Manufacturer:  Polymedco™ 261170
 View more versions of this product
Catalog No. 21-000-113
Add to cart
Description
Description
Available vacuum or nonvacuum and with or without diluent.
Specifications
Specifications
3.8% Sodium Citrate
1000 Box
Non Vacuum ESR Tube without Diluent
Polymedco sediten 10-Minute ESR reader
US Federal and state regulations
Diluent
Clear liquid
5.65