Learn More
Polyoxyethylene bis(amine), M.W. 3,400, Thermo Scientific Chemicals
Description
Polyoxyethylene bis(amine) with HCl, it act as reduction agent; stereo selective allylation of carbonyl compounds; in situ generation of tinenolates for directed aldol reactions. It is used in the preparation of enzyme conjugates, promising drug carrier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Granular Solid |
Solubility Information | Soluble in organic solvents. |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Chemical Name or Material | Polyoxyethylene bis (amine), M.W. 3,400 |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.