Alfa Aesar™ Polyoxyethylene bis(amine), M.W. 8,000

Manufacturer: Alfa Aesar™ 4690303

Quantity 1g

 View more versions of this product

Catalog No. AA4690303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24991-53-5
    Molecular Formula H2NCH2CH2(OCH2CH2)nNH2
    MDL Number MFCD00132004
    Synonym PEG-diamine
    Chemical Name or Material Polyoxyethylene bis (amine), M.W. 8,000

    Specifications

    Physical Form Granular Solid
    Molecular Formula H2NCH2CH2(OCH2CH2)nNH2
    CAS 24991-53-5
    Synonym PEG-diamine
    Quantity 1g
    MDL Number MFCD00132004
    Sensitivity Air Sensitive
    Color White to off-white
    Chemical Name or Material Polyoxyethylene bis (amine), M.W. 8,000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit