Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polypeptide GalNac Transferase 10/GALNT10 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177058MFV500
Description
Polypeptide GalNac Transferase 10/GALNT10 Polyclonal antibody specifically detects Polypeptide GalNac Transferase 10/GALNT10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Polypeptide GalNac Transferase 10/GALNT10 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
DKFZp586H0623, EC 2.4.1.41, FLJ00205, FLJ11715, GalNAc transferase 10, GalNAc-T10, GALNACT10, Polypeptide GalNAc transferase 10, polypeptide N-acetylgalactosaminyltransferase 10, PPGALNACT10, pp-GaNTase 10, PPGANTASE10, Protein-UDP acetylgalactosaminyltransferase 10, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 10, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 10 (GalNAc-T10) | |
Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human GALNT10. The immunogen is located within amino acids 530 - 580 of GALNT10. Amino Acid Squence: KGNQLWKYRKDKTLYHPV | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
55568 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction