Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polypeptide GalNac Transferase 4/GALNT4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Polypeptide GalNac Transferase 4/GALNT4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP162381
|
Novus Biologicals
NBP162381 |
100 μL |
Each for $436.00
|
|
NBP1623820
|
Novus Biologicals
NBP16238120UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Polypeptide GalNac Transferase 4/GALNT4 Polyclonal specifically detects Polypeptide GalNac Transferase 4/GALNT4 in Human samples. It is validated for Western Blot.Specifications
Polypeptide GalNac Transferase 4/GALNT4 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
8693 | |
IgG | |
Affinity Purified | |
67 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q8N4A0 | |
GALNT4 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title