Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Polypropylene, Cap Adapter with Four 3/8 in. Hose Barbs, 120 mm

For Use With: carboys with 120mm cap

Manufacturer:  Justrite™ 12883

Catalog No. 19-830-482

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

  • Interchangeable cap and adapter for top connection flexibility for carboys
  • Autoclavable
  • For quick and easy port changes
  • Two-piece open cap and adapter prevents tube twisting
Specifications

Specifications

Adapter for Carboy Cap with 3/8 in. Hose barb; 4 disconnects
Polypropylene
carboys with 120mm cap
H x Dia.: 3.7 x 4.7 in.
Cap Adapter
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit