Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Polypropylene, Cap Adapter with Three 1/4 in. Hose Barbs, 83 mm

For Use With: carboys with 83mm cap

Supplier:  Justrite™ 12871

Catalog No. 19-830-470

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

  • Interchangeable cap and adapter for top connection flexibility for carboys
  • Autoclavable
  • For quick and easy port changes
  • Two-piece open cap and adapter prevents tube twisting
Specifications

Specifications

Cap Adapter
H x Dia.: 2.8 x 3.2 in.
carboys with 83mm cap
Adapter for Carboy Cap with 1/4 in. Hose barb; 3 disconnects
Polypropylene
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit