Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Polypropylene, Carboy
Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
40 L
75 L
Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Constructed from autoclavable USP Class VI, FDA grade materials
Specifications
Accuracy | ±5% |
Product Type | Carboy |
Capacity (English) | 10.6 gal. |
Closure Material | Polypropylene |
Closure Size | 120 mm |
Capacity (Metric) | 40 L |
Dimensions (L x W x H) | 434 x 249 x 772 mm |
Material | Polypropylene |
For Use With (Application) | For storing and dispensing solvents, buffers, reagents and powders. |
Height (English) | 30.4 in. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction