Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Polypropylene, Carboy with Spigot
For Use With: For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.; Autoclavable; Closure Material: Polypropylene; Color: White
Supplier: Justrite™ 12942
Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Constructed from autoclavable USP Class VI, FDA grade materials
Specifications
±5% | |
Polypropylene | |
40 L | |
Polypropylene | |
For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed. | |
Spigot | |
Rectangular | |
Autoclavable |
10.6 gal. | |
120 mm | |
476 x 249 x 772 mm | |
Carboy with Spigot | |
30.4 in. | |
18.75 in. | |
9.8 in. | |
White |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title