Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Polypropylene Graduated Cylinders

Molded of chemical-resistant, autoclavable polypropylene

 View more versions of this product

Catalog No. S32800K

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Designed for simplicity and ease of use
  • Molded-in graduations will not fade, wash off, or dissolve with chemicals
  • Large, hexagonal base ensures stability
  • Polypropylene construction eliminates meniscus, making reading contents easier and more accurate
  • Available as a set of six, including one each of the 25, 50, 100, 250, 500, and 1000mL size

Specifications

Specifications

Round
1000 mL, 100 mL, 25 mL, 250 mL, 50 mL
140 mm
Polypropylene
DIN 12680 TT, ISO 6706
0.55 in.
Autoclavable
Hexagonal
5.5 in.
Graduated Cylinder Set
±0.2 mL
Class B
1.4 cm
121°C
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit