Learn More
Endicott-Seymour Endicott-Seymour Polypropylene Holders and Dryers [C,P]
Store easily in freezer or refrigerator
Manufacturer:  Endicott-Seymour 237
 View more versions of this product
Catalog No. 14-781-18
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
Store easily in freezer or refrigerator
  • One-piece molded polypropylene support consists of base unit and many individual posts
  • Holds tubes placed open-end-up between posts
  • Dries tubes placed open-end-down over the posts
  • Autoclavable
Specifications
Specifications
Not Autoclavable
Polypropylene
Drying Rack
27.94 cm
10.2 cm
7.62 cm
Tubes (30)
44
22 to 25mm dia. Tubes
11 in.
4 in.
3 in.