Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Polypropylene Lab Coats

These Polypropylene Lab Coats have three pockets and provide economical, lightweight protection against dirt, grime and particulates in non-hazardous environments.

$56.70 - $63.70

Specifications

Closure Type Snaps
Color White
Gender Unisex
Material Polypropylene
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-472-472
View Documents
International Enviroguard™
K2045M
Medium
Case of 30 for $56.70
Add to cart
 
19-472-473
View Documents
International Enviroguard™
K2045L
Large
Case of 30 for $58.40
Add to cart
 
19-472-474
View Documents
International Enviroguard™
K2045XL
X-Large
Case of 30 for $59.20
Add to cart
 
19-472-475
View Documents
International Enviroguard™
K20452XL
2X-Large
Case of 30 for $60.00
Add to cart
 
19-472-476
View Documents
International Enviroguard™
K20453XL
3X-Large
Case of 30 for $61.80
Add to cart
 
19-472-477
View Documents
International Enviroguard™
K20454XL
4X-Large
Case of 30 for $63.70
Add to cart
 
Description

Description

Whether you're working in a full-scale laboratory, a food processing plant, or at an environmental cleanup site, polypropylene lab coats help protect you from dust, dirt, grime, and other dry particulates in non-hazardous environments. Breathable fabric helps prevent heat stress and keeps you cool over long hours. With three spacious pockets, you can carry eyeglass, pens, and other small instruments with you on the go.
  • Low-cost protection against dirt, grime.
  • Three spacious pockets to carry various items and small instruments.
  • Recommended applications include food processing, pharmaceuticals, environmental cleanup, and more.
Specifications

Specifications

Snaps
White
Unisex
Polypropylene
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit