Learn More
Biotix™ Polypropylene Microcentrifuge Tubes
Tubes with superior clarity for visualization of samples.
Manufacturer:  Biotix™ MT0150BCS
 View more versions of this product
Catalog No. 12-111-106
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • DNase, RNase and Endotoxin (pyrogen) free
  • Made from medical-grade virgin polypropylene
Specifications
Specifications
NA
1.5mL
6.38mm
Polypropylene
Sterile
No
Clear
37.8mm
20000 RCF