Learn More
Biotix™ Polypropylene Microcentrifuge Tubes
Tubes with superior clarity for visualization of samples.
Manufacturer:  Biotix™ MT0200R
 View more versions of this product
Catalog No. 12-111-112
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • DNase, RNase and Endotoxin (pyrogen) free
  • Made from medical-grade virgin polypropylene
Specifications
Specifications
Non-sterile
No
Red/Green/Yellow/Blue/Orange
37.9mm
20000 RCF
NA
2mL
6.38mm
Polypropylene