Learn More
Biotix™ Polypropylene PCR Tube
Feature ultra-thin wall that provides excellent thermal transfer with PCR heat blocks.
Manufacturer:  Biotix™ PCT00028C
Catalog No. 12-111-114
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Precision-engineered cap seal prevents amplification errors due to evaporation
  • DNase, RNase and Endotoxin (pyrogen) free
Specifications
Specifications
Not Autoclavable
Non-sterile
Clear
Not Included
8-Strip Tube
0.2 mL