Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynalon Polypropylene Pipets
Easy to read with indelible calibrations.
$20.00 - $158.00
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
S34788C
![]() |
Dynalon
3027650005 |
5 mL |
Each for $20.00
Case of 10 EA for $200.00 |
|
S34788A
![]() |
Dynalon
3027650001 |
1 mL |
Case of 12 for $158.00
|
|
S34788D
![]() |
Dynalon
3027650010 |
10 mL |
Case of 10 for $103.49
|
|
Description
These quality pipets are unbreakable, autoclavable and highly chemical resistant.
- Made of polypropylene
- Completely calibrated “To Deliver” to the tip
- No meniscus; last drop must be blown out
- Note: Exposure to temperatures above 60ºC may cause volume changes.
Specifications
Yes | |
Polypropylene | |
0.1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title