Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand Polypropylene Tubing T-Shaped Connectors
Withstand flows to 20psi.
Description
- Excellent chemical resistance
- Polypropylene
- Autoclavable
- Serrated connecting surfaces to prevent tubing slipping
- Withstands flows to 20 p.s.i.
Specifications
Transparent | |
Autoclavable | |
Polypropylene | |
80.6 mm | |
14 mm | |
0.5 in. | |
5.08 mm |
20 psi | |
0.562 in. | |
3.125 in. | |
Tubing Connector | |
12.7 mm | |
T | |
0.2 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title