Learn More
Brady™ Polypropylene Warning Sign: FLAMMABLE KEEP FLAMES AND HEAT AWAY
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$28.90 - $31.50
Specifications
Color | Red on White |
---|---|
Shape | Rectangular |
Displayed Text | FLAMMABLE KEEP FLAMES AND HEAT AWAY |
Temperature Range (English) | -40°F to 185°F |
Finish | Not Laminated |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-104-889
|
Brady™
116161 |
10 x 7 in. |
Each for $28.90
|
|
|||||
19-104-888
|
Brady™
116160 |
14 x 10 in. |
Each for $31.50
|
|
|||||
Description
- High density polypropylene material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- For indoor and outdoor use
Specifications
Red on White | |
FLAMMABLE KEEP FLAMES AND HEAT AWAY | |
Not Laminated | |
1 Ea. | |
High density polypropylene | |
185°F | |
0.0551 in. |
Rectangular | |
-40°F to 185°F | |
Polypropylene | |
Non-adhesive | |
Corner holes | |
-40°F |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.