Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Polypropylene Watch Glasses Products for Science Education

Allow closer observation of precipitates.

Manufacturer:  Dynalon™ 255235

 View more versions of this product

Catalog No. S34827A



Add to cart

Description

Description

  • Stable base
  • Made of chemical-resistant polypropylene
  • Withstands temperatures to 120°C
  • May also be used as beaker cover

Specifications

Specifications

2.36 in.
Polypropylene
For beakers
≤248°F
6 cm
Used as beaker covers
9.1 mm
≤120°C
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit