Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysialic acid Chimeric Recombinant Rabbit Monoclonal Antibody (735), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA547855

Encompass

Catalog No. PIMA547855


Add to Cart

Description

Description

Specificity: Recognises Polysialic acid (PSA), a carbohydrate polymer attached to the neural cell adhesion molecule (NCAM), which shares molecular epitopes with homopolymers of alpha-2,8-linked sialic acid of Escherichia coli K1.

Polysialic acid Chimeric Recombinant Monoclonal antibody specifically detects Polysialic acid Chimeric in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polysialic acid Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
Rabbit
Affinity chromatography
RUO
Mouse
Liquid
Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
735
Unconjugated
polySia; PSA
Purified murine CR1 protein.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG κ
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit