Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 20, Multi-Compendial, N.F., J.T.Baker™

Click to view available options
Quantity:
20mL
200mL
1L
4L
19L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
Steel Drum
9005-64-5
C26H50O10
522.676
HMFKFHLTUCJZJO-UHFFFAOYSA-N
443314
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
9005-64-5
C26H50O10
522.676
HMFKFHLTUCJZJO-UHFFFAOYSA-N
443314
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
Packaging Steel Drum
Grade Multi-Compendial/NF
Flash Point 148.00°C
Quantity 19L
Specific Gravity 1L = 1.11kg
Chemical Name or Material Polysorbate 20