Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 80, Vegetable, Multi-Compendial, N.F., J.T.Baker™

Manufacturer:  J.T.Baker 450012

Packaging Narrow Mouth Amber Glass Bottle, w/ Septum
Quantity 1L

 View more versions of this product

Catalog No. 02-003-661


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

9005-65-6
Polysorbate 80, Vegetable
Multi-Compendial/NF

Specifications

9005-65-6
148.00°C
1L = 1.08kg
Multi-Compendial/NF
Narrow Mouth Amber Glass Bottle, w/ Septum
1L
Polysorbate 80, Vegetable
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit