Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polystyrene: 1% Divinylbenzene Copolymer Beads 100-200 mesh, ACROS Organics™

Chemical Identifiers

CAS 9003-70-7
MDL Number MFCD00146442
Chemical Name or Material Polystyrene: 1% divinylbenzene copolymer beads, 100-200 mesh
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
AC418081000 SDS Acros Organics
418081000
Glass bottle 100g N/A N/A N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications

Chemical Identifiers

9003-70-7
MFCD00146442
Polystyrene: 1% divinylbenzene copolymer beads, 100-200 mesh

Safety and Handling

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit