Alfa Aesar™ Polystyrene, atactic

Manufacturer: Alfa Aesar™ 4453709

Quantity 10g

 View more versions of this product

Catalog No. AA4453709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9003-53-6
    Formula Weight 125,000 to 250,000
    MDL Number MFCD00084450
    Chemical Name or Material Polystyrene, Atactic

    Specifications

    Formula Weight 125,000 to 250,000
    TSCA TSCA
    Physical Form Beads
    CAS 9003-53-6
    Quantity 10g
    MDL Number MFCD00084450
    Odor Odorless
    Chemical Name or Material Polystyrene, Atactic

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit