Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polystyrene Caution Sign: CONTAINS BENZENE CANCER HAZARD

Improve workplace safety with proper signage. The Brady™ Caution Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

$11.25 - $16.25

Specifications

Adhesive Type Non-adhesive
Color Black on Yellow
Description Laminated Polystyrene
Message CONTAINS BENZENE CANCER HAZARD
Quantity 1 Ea.
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-105-505
View Documents
Brady™
125941
10 x 7 in.
Each for $11.25
Add to cart
 
19-105-508
View Documents
Brady™
125944
14 x 10 in.
Each for $16.25
Add to cart
 
Description

Description

  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Polystyrene signs are thin, pliable, and durable
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
Specifications

Specifications

Non-adhesive
Laminated Polystyrene
1 Ea.
0.0591 in.
Black on Yellow
CONTAINS BENZENE CANCER HAZARD
Rectangular
Polystyrene
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit