Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Polystyrene Danger Sign: FLAMMABLE KEEP FLAMES AND HEAT AWAY
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$18.11 - $26.55
Specifications
Color | Black/Red on White |
---|---|
Message | FLAMMABLE KEEP FLAMES AND HEAT AWAY |
Shape | Rectangular |
Temperature Range (English) | -40°F to 176°F |
Finish | Not Laminated |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-106-922
|
Brady™
25662 |
10 x 7 in. |
Each for $18.11
|
|
19-106-923
|
Brady™
25663 |
14 x 10 in. |
Each for $26.55
|
|
Description
- Polystyrene material is abrasion-resistant and chemical-resistant
- Polystyrene signs are thin, pliable, and durable
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Black/Red on White | |
Rectangular | |
Not Laminated | |
1 Ea. | |
Corner holes | |
-40°F | |
Non-adhesive |
FLAMMABLE KEEP FLAMES AND HEAT AWAY | |
-40°F to 176°F | |
Polystyrene | |
Laminated Polystyrene | |
176°F | |
0.0591 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title