Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Polystyrene Multi-use Test Tubes

Molded from premium grade polystyrene for clarity

Supplier:  Globe Scientific 110409

 View more versions of this product

Catalog No. 22-010-079


Add to Cart

Description

Description

  • Multi-use tubes
  • Can withstand 1500rpm
Specifications

Specifications

Clear
75 mm
250/Bag, 8 Bags/Case
Test Tube
5 mL
Polystyrene
12 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit