Learn More
Argos Technologies Polystyrene Pipetting Reservoir Basins
Designed to work with 8 and 12 channel pipettes. Argos Technologies Polystyrene Pipetting Reservoir Basins allows secure grip and retains shape during handling.
Manufacturer:  Argos Technologies B3110100NS
 View more versions of this product
Catalog No. 03-391-543
  Check Availability
Add to cart
Description
Description
  • Angled sides and narrow V-shaped bottom ensures every last drop of reagents can be aspirated
  • Certified RNase, DNase and pyrogen free
  • Graduations on inner walls
  • Larger spout on each reservoir to facilitate spill-free pouring
  • Packaged in easy open tear strip bags
Specifications
Specifications
Non-sterile
Polystyrene
100
Pipetting Reservoir
100mL
Bulk pack
White