Learn More
Brady™ Polystyrene Sign: HIGH VOLTAGE. CONTACT COULD RESULT IN SERIOUS SHOCK OR DEATH.
Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 144394
 View more versions of this product
Catalog No. 19-109-563
Add to cart
Description
Description
  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Polystyrene signs are thin, pliable and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Orange, Yellow, Black on White
Rectangle
HIGH VOLTAGE. CONTACT COULD RESULT IN SERIOUS SHOCK OR DEATH.
PS (Polystyrene)
14 in. x 10 in. x 0.0591 in.