Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Brady™ Polystyrene Sign: HEARING PROTECTION MUST BE WORN IN THIS AREA
Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 140802
Catalog No. 19-111-604
Description
Description
  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Polystyrene signs are thin, pliable and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Blue on White
Rectangle
HEARING PROTECTION MUST BE WORN IN THIS AREA
PS (Polystyrene)
23.875 in. x 6 in. x 0.0591 in.