Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Polystyrene Sign: HIGH VOLTAGE. CONTACT WILL RESULT IN SERIOUS SHOCK OR DEATH.
Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$21.00 - $30.75
Specifications
Material | Polystyrene |
---|---|
Shape | Rectangular |
Displayed Text | HIGH VOLTAGE. CONTACT WILL RESULT IN SERIOUS SHOCK OR DEATH. |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-109-554
|
Brady™
144385 |
14 x 10 in. |
Each for $30.75
|
|
|||||
19-109-557
|
Brady™
144388 |
10 x 7 in. |
Each for $21.00
|
|
|||||
Description
- Polystyrene material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Polystyrene signs are thin, pliable and durable
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Polystyrene | |
Rectangular | |
HIGH VOLTAGE. CONTACT WILL RESULT IN SERIOUS SHOCK OR DEATH. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title