Learn More
Brady™ Polystyrene Sign: STAY CLEAR. AUTOMATIC START EQUIPMENT.
Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 143926
 View more versions of this product
Catalog No. 19-110-767
Add to cart
Description
Description
  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Polystyrene signs are thin, pliable and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Red, Yellow, Black on White
Rectangle
STAY CLEAR. AUTOMATIC START EQUIPMENT.
PS (Polystyrene)
10 in. x 7 in. x 0.0591 in.