Polystyrene standard, M.W. 30,000, Mw/Mn 1.06, Alfa Aesar™

Manufacturer: Alfa Aesar 4193006

Quantity 5g

 View more versions of this product

Catalog No. AA4193006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9003-53-6
    Physical Form Powder
    MDL Number MFCD00084450
    Chemical Name or Material Polystyrene standard, M.W. 30, 000, Mw/Mn 1.06

    Specifications

    Physical Form Powder
    Quantity 5g
    Color White
    CAS 9003-53-6
    Ignition Point 427°C (801°F)
    Odor Odorless
    Appearance White powder
    MDL Number MFCD00084450
    Chemical Name or Material Polystyrene standard, M.W. 30, 000, Mw/Mn 1.06

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit