Learn More
Brady™ Polystyrene Warning Sign: DO NOT ENTER HAZARDOUS AREA AUTHORIZED PERSONS ONLY
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$18.90 - $28.35
Specifications
Color | Red on White |
---|---|
Shape | Rectangular |
Displayed Text | DO NOT ENTER HAZARDOUS AREA AUTHORIZED PERSONS ONLY |
Temperature Range (English) | -40°F to 176°F |
Finish | Not Laminated |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-105-718
|
Brady™
126205 |
10 x 7 in. |
Each for $18.90
|
|
|||||
19-105-721
|
Brady™
126208 |
14 x 10 in. |
Each for $28.35
|
|
|||||
Description
- Polystyrene material is abrasion-resistant and chemical-resistant
- Polystyrene signs are thin, pliable, and durable
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Red on White | |
DO NOT ENTER HAZARDOUS AREA AUTHORIZED PERSONS ONLY | |
Not Laminated | |
1 Ea. | |
Laminated Polystyrene | |
176°F | |
0.0591 in. |
Rectangular | |
-40°F to 176°F | |
Polystyrene | |
Non-adhesive | |
Corner holes | |
-40°F |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.