Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polystyrene Warning Sign: LEAD WORK AREA. POISON. NO SMOKING OR EATING.

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$11.25 - $16.25

Specifications

Color Black/orange/red on white
Material PS (Polystyrene)
Message LEAD WORK AREA. POISON. NO SMOKING OR EATING.
Min. Temperature (English) -40°F
Quantity 1/Ea.
View More Specs

Products 2
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
19-106-619 View Documents Brady™
145160
14 in. x 10 in. x 0.0591 in. Each for $16.25
Add to cart
 
19-106-622 View Documents Brady™
145163
10 in. x 7 in. x 0.0591 in. Each for $11.25
Add to cart
 
Description

Description

  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Polystyrene signs are thin, pliable, and durable
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
Specifications

Specifications

Black/orange/red on white
LEAD WORK AREA. POISON. NO SMOKING OR EATING.
1/Ea.
14 in.
Rectangle
PS (Polystyrene)
-40°F
Corner holes
10 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit