Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polystyrene Warning Sign: LP GAS STORAGE. NO SMOKING OR OPEN FLAMES WITHIN 50 FT.

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$11.25 - $16.25

Specifications

Color Black/red on white
Message LP GAS STORAGE. NO SMOKING OR OPEN FLAMES WITHIN 50 FT.
Quantity 1/Ea.
Mounting Type Corner holes
Width (English) 14 in.
View More Specs

Products 2
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
19-106-635 View Documents Brady™
145176
14 in. x 10 in. x 0.0591 in. Each for $16.25
Add to cart
 
19-106-636 View Documents Brady™
145177
10 in. x 7 in. x 0.0591 in. Each for $11.25
Add to cart
 
Description

Description

  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications

Specifications

Black/red on white
1/Ea.
14 in.
Rectangle
LP GAS STORAGE. NO SMOKING OR OPEN FLAMES WITHIN 50 FT.
Corner holes
10 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit