Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polystyrene Warning Sign: STORAGE AREA FLAMMABLE MATERIAL

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$19.43 - $28.35

Specifications

Color Black/Red on White
Message STORAGE AREA FLAMMABLE MATERIAL
Shape Rectangular
Temperature Range (English) -40°F to 176°F
Finish Not Laminated
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-106-925
View Documents
Brady™
25678
10 x 7 in.
Each for $19.43
Add to Cart
 
19-106-926
View Documents
Brady™
25679
14 x 10 in.
Each for $28.35
Add to Cart
 
Description

Description

  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Polystyrene signs are thin, pliable, and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications

Specifications

Black/Red on White
Rectangular
Not Laminated
1 Ea.
Corner holes
-40°F
Non-adhesive
STORAGE AREA FLAMMABLE MATERIAL
-40°F to 176°F
Polystyrene
Laminated Polystyrene
176°F
0.0591 in.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit