Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysulfone resin, sec. stand., typical M.W.67000, typical M.N.20000, Acros Organics

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
25135-51-7
C27H26O6S
478.559
MFCD00134387
ZUZZUJNBGCVPLQ-UHFFFAOYSA-N
16211415
4-[2-(4-hydroxyphenyl)propan-2-yl]phenol;4-(4-hydroxyphenyl)sulfonylphenol
CC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O.C1=CC(=CC=C1O)S(=O)(=O)C2=CC=C(C=C2)O
25135-51-7
C27H26O6S
478.559
MFCD00134387
ZUZZUJNBGCVPLQ-UHFFFAOYSA-N
16211415
4-[2-(4-hydroxyphenyl)propan-2-yl]phenol;4-(4-hydroxyphenyl)sulfonylphenol
CC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O.C1=CC(=CC=C1O)S(=O)(=O)C2=CC=C(C=C2)O
Density 1.2400g/mL
Physical Form Pellets
Infrared Spectrum Authentic
Packaging Glass bottle
Color Undesignated
Quantity 25g
Chemical Name or Material Polysulfone resin