Alfa Aesar™ Polytetrafluoroethylene, 6-10 micron

Manufacturer: Alfa Aesar™ 4418418

Quantity 50g

 View more versions of this product

Catalog No. AA4418418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9002-84-0
    Molecular Formula −(CF2CF2-)n
    MDL Number MFCD00084455
    Merck Index 14,7583
    Chemical Name or Material Polytetrafluoroethylene

    Specifications

    MDL Number MFCD00084455
    CAS 9002-84-0
    RTECS Number KX4025000
    TSCA TSCA
    Physical Form Powder
    Molecular Formula −(CF2CF2-)n
    Merck Index 14,7583
    Particle Size 6 to 10 Micron
    Quantity 50g
    Chemical Name or Material Polytetrafluoroethylene

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit