Learn More
InterLab™ Supply PolyTox™ Rapid Toxicity Test Vials
30 Minute rapid test for measuring toxicity of wastewater to biological systems
Manufacturer:  InterLab™ Supply P410
Catalog No. 13-297-202
Add to cart
Description
Description
A blend of specialized microbial cultures in an easy-to-use kit provides a simple and cost-effective test.
  • EPA recommended
  • No expensive instrumentation required
  • Non-pathogenic
  • Free of nitrifying microorganisms
  • Reliable results in less than 30 minutes
  • Each kit contains 20 vials
Specifications
Specifications
EPA
Measuring toxicity of wastewater to biological wastewater treatment systems
30 min.
PolyTox™ Rapid Biological Toxicity Screening Test
20 vials, each vial holds 8 oz.
Safety and Handling
Safety and Handling
ShelfLife : 3 to 6 Months