Learn More
Kinematica™ Polytron™ 10-35GT, 3100D, 45-80GT & 6100D Generator/Probes
Polytron™ dispersing aggregates are available in different designs, like rotor with knives for pre-cutting, rotor/stator with several teeth rows for fine or double emulsions, mechanical seals for vacuum and pressure applications, and more.
$1941.00 - $4100.00
Specifications
For Use With (Equipment) Polytron™ 10-35 GT,3100 D, 45-80 GT, 6100 D Homogenizers
Material 316L Stainless Steel
View More Specs
Products 17
Catalog Number Mfr. No. Bottom Type Diameter (Metric) Length (Metric) Shaft Model Price Quantity & Availability  
Catalog Number Mfr. No. Bottom Type Diameter (Metric) Length (Metric) Shaft Model Price Quantity & Availability  
08-451-019
View Documents
Kinematica™
11030297
Sawtooth 5 mm 78 mm PT-DA 05/2 EC-F078
Each for $1,941.00
Add to cart
08-451-020
View Documents
Kinematica™
11030298
Sawtooth 7 mm 101 mm PT-DA 07/2 EC-F101
Each for $2,110.00
Add to cart
08-451-021
View Documents
Kinematica™
11030299
Sawtooth 12 mm 154 mm PT-DA 12 / 2 EC-F154
Each for $2,190.00
Add to cart
08-451-022
View Documents
Kinematica™
11030300
Knive Rotors 12 mm 154 mm PT-DA 12/2 MEC-F154
Each for $2,360.00
Add to cart
08-451-023
View Documents
Kinematica™
11030301
W Stator Teeth 12 mm 154 mm PT-DA 12/2 WEC-F154
Each for $2,630.00
Add to cart
08-451-024
View Documents
Kinematica™
11030302
Sawtooth 20 mm 193 mm PT-DA 20/2 EC-F193
Each for $2,440.00
Add to cart
08-451-025
View Documents
Kinematica™
11030303
Knive Rotors 20 mm 193 mm PT-DA 20/2 MEC-F193
Each for $2,650.00
Add to cart
08-451-026
View Documents
Kinematica™
11030304
W Stator Teeth 20 mm 193 mm PT-DA 20/2 WEC-F193
Each for $2,920.00
Add to cart
08-451-027
View Documents
Kinematica™
11030305
Sawtooth 30 mm 250 mm PT-DA 30/2 EC-F250
Each for $3,080.00
Add to cart
08-451-030
View Documents
Kinematica™
11030309
W Stator Teeth 30 mm 250 mm PT-DA 30/2 WEC-F250
Each for $3,770.00
Add to cart
08-451-031
View Documents
Kinematica™
11030311
Sawtooth 30 mm 250 mm PT-DA 30/4 EC-F250
Each for $3,940.00
Add to cart
08-451-032
View Documents
Kinematica™
11030313
Sawtooth 36 mm 250 mm PT-DA 36/2 EC-F250
Each for $3,280.00
Add to cart
08-451-033
View Documents
Kinematica™
11030314
Biotrona™ 36 mm 250 mm PT-DA 36/BEC-F250 (BIOTRONA)
Each for $3,590.00
Add to cart
08-451-034
View Documents
Kinematica™
11030315
Knive Rotors 36 mm 250 mm PT-DA 36/2 MEC-F250
Each for $3,580.00
Add to cart
08-451-036
View Documents
Kinematica™
11030319
W Stator Teeth 36 mm 250 mm PT-DA 36/2 WEC-F250
Each for $4,100.00
Add to cart
08-451-035
View Documents
Kinematica™
11030317
Sawtooth 36 mm 250 mm PT-DA 36/4 EC-F250
Each for $4,100.00
Add to cart
08-451-016
View Documents
Kinematica™
11030264
Sawtooth 25 mm 192 mm PT-DA 25/2 EC-E192
Each for $2,760.00
Add to cart
Description
Description
  • /2 Design: Universal geometry for emulsifying, suspending, dispersing, and extracting samples
  • W Design: Used for fibrous or stringy samples
  • M Design: Rotor with pre-cutting knives, ideal for dispersing samples larger than rotors diameter
  • Biotrona Design: High-turbulence mixing head for highly viscous samples
  • Autoclave Safe
  • Includes Instruction Manual and Tool for easy dissambly
  • Click and Go Coupling System allows for one hand assembly
  • Warranty: 2 years
Specifications
Specifications
Polytron™ 10-35 GT,3100 D, 45-80 GT, 6100 D Homogenizers
316L Stainless Steel