Learn More
Fisherbrand™ Polyurethane Chair - Desk Height with Medium Back, Seat Tilt, and Casters in Polyurethane
Fisherbrand™ Polyurethane Chairs with Medium Back are height adjustable for 31 in. work surface and feature ergonomic adjustments and self-braking casters. Self-skinning chemical, soil, and puncture resistant polyurethane makes this ideal for any environment.
$353.00 - $679.50
Specifications
Adjustable Yes
Base Material Nylon Reinforced Fiberglass
Material Polyurethane
Footrest No
With Armrest No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-538
View Documents
Blue
Each for $353.00
Add to cart
14-360-444
View Documents
Black
Each for $679.50
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 16.25-21.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Polyurethane
No
Yes
Desk Height
Nylon Reinforced Fiberglass
No
Yes
Yes